Learn More
Chemglass Life Sciences Chemglass O-ring, Simriz 486 FFKM (perfluoro), Size 219 - O-ring for laboratory glassware seals
SDP
Catalog No. 502743483
Encompass_Preferred
Chemglass CG-309-Z-219 is a white, high-purity perfluoro (FFKM) O-ring made from Simriz 486 material. It provides broad chemical resistance and high temperature stability for demanding laboratory, pharmaceutical, and semiconductor sealing applications.
  • High chemical and solvent resistance suitable for aggressive process chemistries.
  • White, high-purity perfluoroelastomer suitable for cleanroom and pharmaceutical use.
  • USP Class VI certified for biocompatibility.
  • Heat resistant to approximately 220°C for high-temperature applications.
  • Suitable for seals in laboratory glassware, reaction vessels, and vacuum systems.
Catalog No. 50-274-3483 Supplier Chemglass Life Sciences Supplier No. CG309Z219
Chemglass CG-309-Z-219 is a white, high-purity perfluoro (FFKM) O-ring made from Simriz 486 material. It provides broad chemical resistance and high temperature stability for demanding laboratory, pharmaceutical, and semiconductor sealing applications.
  • High chemical and solvent resistance suitable for aggressive process chemistries.
  • White, high-purity perfluoroelastomer suitable for cleanroom and pharmaceutical use.
  • USP Class VI certified for biocompatibility.
  • Heat resistant to approximately 220°C for high-temperature applications.
  • Suitable for seals in laboratory glassware, reaction vessels, and vacuum systems.