Learn More
Chemglass Life Sciences Chemglass Perfluoro O-Ring (FFKM) AS568 #220, White, Class VI
SDP
Catalog No. 502743482
Encompass_Preferred
Perfluoro (FFKM) O-ring manufactured from Simriz 486 material, designed for high chemical and thermal resistance in laboratory vacuum and O-ring joint applications.
  • Perfluoroelastomer (FFKM) material.
  • Service temperature: -7 °C to 230 °C.
  • White color, 75 Shore A durometer.
  • USP Class VI compliant.
  • AS568 dash #220 size for standard O-ring joints.
  • Suitable for vacuum traps and O-ring joints.
Catalog No. 50-274-3482 Supplier Chemglass Life Sciences Supplier No. CG309Z220
Perfluoro (FFKM) O-ring manufactured from Simriz 486 material, designed for high chemical and thermal resistance in laboratory vacuum and O-ring joint applications.
  • Perfluoroelastomer (FFKM) material.
  • Service temperature: -7 °C to 230 °C.
  • White color, 75 Shore A durometer.
  • USP Class VI compliant.
  • AS568 dash #220 size for standard O-ring joints.
  • Suitable for vacuum traps and O-ring joints.