Learn More
Chemglass Life Sciences O-Ring #222, Simriz 486 FFKM (Perfluoro), USP Class VI - Chemglass CG-309-Z-222
SDP
Catalog No. 502743481
Encompass_Preferred
O-Ring #222 made from Simriz 486 FFKM (perfluoro) is a high-purity sealing component engineered for aggressive chemical environments and elevated temperatures. It combines elastomer resilience with chemical resistance comparable to PTFE and is manufactured in accredited clean-room facilities for critical process use.
  • High chemical and solvent resistance.
  • Wide temperature tolerance: -7°C to +230°C.
  • USP Class VI certified for biocompatibility.
  • Hardness: Shore A 75 for reliable sealing performance.
  • Tensile strength approximately 2620 psi for durability.
  • Manufactured in ISO-certified clean rooms for purity.
  • Sold individually as a single O-ring.
Catalog No. 50-274-3481 Supplier Chemglass Life Sciences Supplier No. CG309Z222
O-Ring #222 made from Simriz 486 FFKM (perfluoro) is a high-purity sealing component engineered for aggressive chemical environments and elevated temperatures. It combines elastomer resilience with chemical resistance comparable to PTFE and is manufactured in accredited clean-room facilities for critical process use.
  • High chemical and solvent resistance.
  • Wide temperature tolerance: -7°C to +230°C.
  • USP Class VI certified for biocompatibility.
  • Hardness: Shore A 75 for reliable sealing performance.
  • Tensile strength approximately 2620 psi for durability.
  • Manufactured in ISO-certified clean rooms for purity.
  • Sold individually as a single O-ring.