Learn More
Chemglass Life Sciences Chemglass O-Ring, Simriz 486 FFKM (Perfluoro), Size #223, White, USP Class VI
SDP
Catalog No. 502743480
Encompass_Preferred
Chemglass supplies a white, USP Class VI perfluoro (FFKM) O-ring in size #223 made from a high-purity perfluoroelastomer. The component is designed for demanding chemical environments and elevated temperatures, and is manufactured in certified cleanroom facilities.
  • High chemical resistance suitable for aggressive process chemistries.
  • Wide service temperature range: -7 °C to 230 °C.
  • USP Class VI certification for biocompatibility.
  • Hardness approximately 75 Shore A and tensile strength ≈ 2,620 psi.
  • White color for high-purity and contamination-sensitive applications.
  • Produced in cleanroom facilities with ISO/TS 16949, ISO 9001, and AS9100 certifications.
Catalog No. 50-274-3480 Supplier Chemglass Life Sciences Supplier No. CG309Z223
Chemglass supplies a white, USP Class VI perfluoro (FFKM) O-ring in size #223 made from a high-purity perfluoroelastomer. The component is designed for demanding chemical environments and elevated temperatures, and is manufactured in certified cleanroom facilities.
  • High chemical resistance suitable for aggressive process chemistries.
  • Wide service temperature range: -7 °C to 230 °C.
  • USP Class VI certification for biocompatibility.
  • Hardness approximately 75 Shore A and tensile strength ≈ 2,620 psi.
  • White color for high-purity and contamination-sensitive applications.
  • Produced in cleanroom facilities with ISO/TS 16949, ISO 9001, and AS9100 certifications.