Learn More
Chemglass Life Sciences O-ring AS568-224 - Simriz 486 FFKM, 75 Shore A, white perfluoroelastomer seal (chemical/process equipment)
SDP
Catalog No. 502743479
Encompass_Preferred
This perfluoroelastomer (FFKM) O-ring in AS568-224 size is formulated for demanding chemical and semiconductor process environments. It delivers high temperature stability, broad chemical resistance, low particulation, and cleanroom suitability.
  • High chemical resistance to aggressive solvents and acids.
  • Wide operating temperature range up to approximately 230 °C.
  • White, carbon-black free compound suitable for clean environments.
  • 75 Shore A hardness for reliable sealing performance.
  • Low particulation and plasma resistance for semiconductor applications.
  • USP Class VI compatibility for pharmaceutical and biotech uses.
Catalog No. 50-274-3479 Supplier Chemglass Life Sciences Supplier No. CG309Z224
This perfluoroelastomer (FFKM) O-ring in AS568-224 size is formulated for demanding chemical and semiconductor process environments. It delivers high temperature stability, broad chemical resistance, low particulation, and cleanroom suitability.
  • High chemical resistance to aggressive solvents and acids.
  • Wide operating temperature range up to approximately 230 °C.
  • White, carbon-black free compound suitable for clean environments.
  • 75 Shore A hardness for reliable sealing performance.
  • Low particulation and plasma resistance for semiconductor applications.
  • USP Class VI compatibility for pharmaceutical and biotech uses.