Learn More
Chemglass Life Sciences Simriz SZ486 FFKM O-Ring - AS568-225 (White, 75 Shore A)
SDP
Catalog No. 502743478
Encompass_Preferred
Simriz SZ486 perfluoroelastomer (FFKM) O-ring sized to AS568-225. This white, carbon-black free FFKM compound is formulated for broad chemical resistance, thermal stability, and low particulate performance required in pharmaceutical, semiconductor, and biotech sealing applications.
  • Perfluoroelastomer material providing broad chemical resistance and thermal stability.
  • White, carbon-black free formulation for reduced particulate and contamination.
  • 75 Shore A durometer for reliable compression set and sealing performance.
  • Wide operating temperature range suitable for steam and plasma processes.
  • Suitable for high-purity and aggressive chemical handling in pharmaceutical and semiconductor environments.
Catalog No. 50-274-3478 Supplier Chemglass Life Sciences Supplier No. CG309Z225
Simriz SZ486 perfluoroelastomer (FFKM) O-ring sized to AS568-225. This white, carbon-black free FFKM compound is formulated for broad chemical resistance, thermal stability, and low particulate performance required in pharmaceutical, semiconductor, and biotech sealing applications.
  • Perfluoroelastomer material providing broad chemical resistance and thermal stability.
  • White, carbon-black free formulation for reduced particulate and contamination.
  • 75 Shore A durometer for reliable compression set and sealing performance.
  • Wide operating temperature range suitable for steam and plasma processes.
  • Suitable for high-purity and aggressive chemical handling in pharmaceutical and semiconductor environments.