Learn More
Chemglass Life Sciences Chemglass Perfluoro O-ring (Simriz 486 FFKM), Size #226
SDP
Catalog No. 502743477
Encompass_Preferred
Perfluoro (FFKM) O-ring made from Simriz 486 perfluoroelastomer, designed for high chemical resistance and thermal stability. Suitable for laboratory vacuum traps, O-ring joints, and aggressive chemical environments that require high-purity seals.
  • Perfluoroelastomer (FFKM) material offering broad chemical resistance.
  • White color, 75 durometer for sealing performance.
  • Size number 226 (AS568-226) for standard O-ring grooves.
  • Designed for high-temperature and aggressive chemical environments.
  • Sold individually (each).
Catalog No. 50-274-3477 Supplier Chemglass Life Sciences Supplier No. CG309Z226
Perfluoro (FFKM) O-ring made from Simriz 486 perfluoroelastomer, designed for high chemical resistance and thermal stability. Suitable for laboratory vacuum traps, O-ring joints, and aggressive chemical environments that require high-purity seals.
  • Perfluoroelastomer (FFKM) material offering broad chemical resistance.
  • White color, 75 durometer for sealing performance.
  • Size number 226 (AS568-226) for standard O-ring grooves.
  • Designed for high-temperature and aggressive chemical environments.
  • Sold individually (each).