Learn More
Chemglass Life Sciences O-Ring Simriz 486 FFKM (AS568-239) - Chemglass CG-309-Z-239
SDP
Catalog No. 502743476
Encompass_Preferred
Chemglass CG-309-Z-239 is an AS568-239 O-ring manufactured from Simriz 486 perfluoroelastomer (FFKM) for high-purity, high-temperature sealing in aggressive dry process chemistries such as reactive plasmas. It is designed for use in semiconductor, pharmaceutical, and biotech equipment where chemical resistance and cleanliness are required.
  • Perfluoroelastomer compound provides broad chemical resistance.
  • White color aids visual cleanliness in controlled environments.
  • Certified to USP Class VI for biocompatibility.
  • Temperature range suitable for high-temperature applications: -7°C to 230°C.
  • Sized to AS568-239 for standardized O-ring fittings.
Catalog No. 50-274-3476 Supplier Chemglass Life Sciences Supplier No. CG309Z239
Chemglass CG-309-Z-239 is an AS568-239 O-ring manufactured from Simriz 486 perfluoroelastomer (FFKM) for high-purity, high-temperature sealing in aggressive dry process chemistries such as reactive plasmas. It is designed for use in semiconductor, pharmaceutical, and biotech equipment where chemical resistance and cleanliness are required.
  • Perfluoroelastomer compound provides broad chemical resistance.
  • White color aids visual cleanliness in controlled environments.
  • Certified to USP Class VI for biocompatibility.
  • Temperature range suitable for high-temperature applications: -7°C to 230°C.
  • Sized to AS568-239 for standardized O-ring fittings.