Learn More
Chemglass Life Sciences Chemglass O-Ring #243, Simriz 486 FFKM (White), USP Class VI
SDP
Catalog No. 502743475
Encompass_Preferred
Chemglass O-Ring #243 is molded from Simriz 486 perfluoroelastomer (FFKM) and supplied white with USP Class VI certification. It combines elastomer resilience with chemical resistance approaching PTFE, making it suitable for aggressive dry-process chemistries and reactive plasma environments.
  • Provides broad chemical resistance to aggressive solvents and process chemistries.
  • Wide operating temperature range from -7°C to +230°C.
  • Hardness: Shore A 75.
  • Tensile strength: 2620 psi.
  • Certified to USP Class VI for high-purity applications.
  • Sold individually (each).
Catalog No. 50-274-3475 Supplier Chemglass Life Sciences Supplier No. CG309Z243
Chemglass O-Ring #243 is molded from Simriz 486 perfluoroelastomer (FFKM) and supplied white with USP Class VI certification. It combines elastomer resilience with chemical resistance approaching PTFE, making it suitable for aggressive dry-process chemistries and reactive plasma environments.
  • Provides broad chemical resistance to aggressive solvents and process chemistries.
  • Wide operating temperature range from -7°C to +230°C.
  • Hardness: Shore A 75.
  • Tensile strength: 2620 psi.
  • Certified to USP Class VI for high-purity applications.
  • Sold individually (each).