Learn More
Ace Glass, Inc. O-RING 3 110 O-RING 3 110
SDP
Catalog No. NC0055638
Encompass_Preferred
Size 110 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. NC0055638 Supplier Ace Glass, Inc. Supplier No. 7855816
Size 110 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.