Learn More
Chemglass Life Sciences Chemglass O-ring, Simriz 486 FFKM (Perfluoro), white, Class VI - O-ring seal for lab glassware and process equipment
SDP
Catalog No. 502743474
Encompass_Preferred
Perfluoro (FFKM) O-ring formulated from Simriz 486 and stocked by Chemglass for seals requiring exceptional chemical resistance and thermal stability. White, Class VI material suitable for laboratory and process sealing applications.
  • Perfluoroelastomer (FFKM) construction.
  • White color, 75 durometer.
  • Class VI material.
  • Effective service temperature: -7 to 230 °C.
  • Broad chemical compatibility with acids, bases, alcohols/ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Suitable for O-ring joints, vacuum traps, reactors, and other laboratory glassware seals.
  • Designed for high-purity and high-temperature sealing applications.
Catalog No. 50-274-3474 Supplier Chemglass Life Sciences Supplier No. CG309Z326
Perfluoro (FFKM) O-ring formulated from Simriz 486 and stocked by Chemglass for seals requiring exceptional chemical resistance and thermal stability. White, Class VI material suitable for laboratory and process sealing applications.
  • Perfluoroelastomer (FFKM) construction.
  • White color, 75 durometer.
  • Class VI material.
  • Effective service temperature: -7 to 230 °C.
  • Broad chemical compatibility with acids, bases, alcohols/ethers, hydrocarbons, esters, ketones, amines, and chlorocarbon solvents.
  • Suitable for O-ring joints, vacuum traps, reactors, and other laboratory glassware seals.
  • Designed for high-purity and high-temperature sealing applications.