Learn More
Chemglass Life Sciences O-Ring, Simriz 486 FFKM Perfluoro, White, Class VI, Size #331
SDP
Catalog No. 502743473
Encompass_Preferred
Chemglass CG-309-Z-331 is a perfluoroelastomer O-ring made from Simriz 486 FFKM, sized #331 for laboratory and process sealing applications. The component is white, Class VI, and designed to resist a wide range of aggressive chemicals while operating across a broad temperature range.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • White color suitable for high-purity environments.
  • Class VI material for biocompatibility and pharmaceutical use.
  • Stocked at 75 durometer.
  • Service temperature range: -7 to 230 °C.
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.
Catalog No. 50-274-3473 Supplier Chemglass Life Sciences Supplier No. CG309Z331
Chemglass CG-309-Z-331 is a perfluoroelastomer O-ring made from Simriz 486 FFKM, sized #331 for laboratory and process sealing applications. The component is white, Class VI, and designed to resist a wide range of aggressive chemicals while operating across a broad temperature range.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • White color suitable for high-purity environments.
  • Class VI material for biocompatibility and pharmaceutical use.
  • Stocked at 75 durometer.
  • Service temperature range: -7 to 230 °C.
  • Compatible with acids, inorganic bases, alcohols/ethers, aromatic hydrocarbons, esters, aldehydes/ketones, aliphatic hydrocarbons, amines, and chlorocarbon solvents.