Learn More
Perkin Elmer US LLC Internal Injector Support Adapter O-Ring, 6.07 mm I.D., for ELAN/NexION 300/350
Catalog No. NC2061127
Encompass_Preferred
This Injector Support Adapter O-Ring, 6.07 mm I.D. and 1.78 mm width, is for use with ELAN and NexION 300/350 ICP-MS series instruments. It is designed for use with both the Twist Type Torch Injector Support Adapter and the Cassette Type Torch Injector Support Adapter. This PTFE O-ring is included in the ELAN Cassette O-Ring Kit for Injector Support for ELAN (N8120100), Injector Adapter Kit for NexION 300/350 (N8140527),and the HF Sample Introduction Kit for NexION 300/350 (N8140507). This O-ring can also be used for the Baffled Quartz Cyclonic Spray Chamber for NexION 300/350 (N8145013) and Baffled Quartz Cyclonic Spray Chamber for ELAN DRCs (WE025221).
Catalog No. NC2061127 Supplier Perkin Elmer US LLC Supplier No. 09210011
This Injector Support Adapter O-Ring, 6.07 mm I.D. and 1.78 mm width, is for use with ELAN and NexION 300/350 ICP-MS series instruments. It is designed for use with both the Twist Type Torch Injector Support Adapter and the Cassette Type Torch Injector Support Adapter. This PTFE O-ring is included in the ELAN Cassette O-Ring Kit for Injector Support for ELAN (N8120100), Injector Adapter Kit for NexION 300/350 (N8140527),and the HF Sample Introduction Kit for NexION 300/350 (N8140507). This O-ring can also be used for the Baffled Quartz Cyclonic Spray Chamber for NexION 300/350 (N8145013) and Baffled Quartz Cyclonic Spray Chamber for ELAN DRCs (WE025221).