Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ O-Ring

Catalog No. XX4002516 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
25mm Gas Line Filter Holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
XX4002516 25mm Gas Line Filter Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XX4002516 Supplier MilliporeSigma™ Supplier No. XX4002516
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

For Use With (Application) Viton 2-021
For Use With (Equipment) 25mm Gas Line Filter Holder
Quantity 5 Pack
Product Type O-Ring
Material Fluoroelastomer
Diameter (Metric) Outside 23.52 mm
Size 2-021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.