Learn More
Perkin Elmer US LLC External Injector Support Adapter O-Ring, 9.25 mm I.D., for ELAN/NexION 300/350
Catalog No. NC2061128
Encompass_Preferred
This Injector Support Adapter O-Ring, 9.25 mm I.D. and 1.78 mm width, is for use with ELAN and NexION 300/350 ICP-MS series instruments. It is designed for use with both the Twist Type Torch Injector Support Adapter and the Cassette Type Torch Injector Support Adapter. Two of these PTFE O-rings are included in the ELAN Cassette O-Ring Kit for Injector Support for ELAN (N8120100). One is also included in the HF Sample Introduction Kit for NexION 300/350 (N8140507).
Catalog No. NC2061128 Supplier Perkin Elmer US LLC Supplier No. 09210012
This Injector Support Adapter O-Ring, 9.25 mm I.D. and 1.78 mm width, is for use with ELAN and NexION 300/350 ICP-MS series instruments. It is designed for use with both the Twist Type Torch Injector Support Adapter and the Cassette Type Torch Injector Support Adapter. Two of these PTFE O-rings are included in the ELAN Cassette O-Ring Kit for Injector Support for ELAN (N8120100). One is also included in the HF Sample Introduction Kit for NexION 300/350 (N8140507).