Learn More
McMaster-Carr High-Temperature High-Purity Silicone O-Ring, 3/32 Fractional Width, Dash Number 110, 50/pk
Catalog No. NC1734755
Encompass
Even in temperature up to 400° F, these silicone O-rings will bounce back after compression. They have a round profile and are suitable for a wide range of static and dynamic sealing applications. All are sized to the SAE standard AS568.
Catalog No. NC1734755 Supplier McMaster-Carr Supplier No. 9396K23
Even in temperature up to 400° F, these silicone O-rings will bounce back after compression. They have a round profile and are suitable for a wide range of static and dynamic sealing applications. All are sized to the SAE standard AS568.