Learn More
Waters Corp Waters O-Ring, Kalrez, AS007, 0.240 mm x 0.063 mm
Catalog No. 50818843 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Waters O-Ring is designed for critical sealing applications, crafted from Kalrez for exceptional chemical resistance and high-temperature performance.
  • Crafted from Kalrez material for superior chemical compatibility.
  • Designed for reliable sealing in demanding environments.
  • Suitable for ion block assembly.
  • Precision dimensions of 0.240 mm x 0.063 mm.
Catalog No. 50-818-843 Supplier Waters Corp Supplier No. 700000903
This Waters O-Ring is designed for critical sealing applications, crafted from Kalrez for exceptional chemical resistance and high-temperature performance.
  • Crafted from Kalrez material for superior chemical compatibility.
  • Designed for reliable sealing in demanding environments.
  • Suitable for ion block assembly.
  • Precision dimensions of 0.240 mm x 0.063 mm.