Learn More
Waters Corp O-Ring, AS011, 5/pk
Supplier:  Waters Corp 700001460
Encompass_Preferred
The AS011 Viton O-Ring is used on the following Mass Spectrometers: GCT, LCT Premier, LCT Premier XE, MALDI LR, Q-Tof Premier, Quattro Premier, Quattro Premier XE, SYNAPT HDMS, SYNAPT MS, and MALDI SYNAPT HDMS/MS Systems.
Catalog No. 50-792-581