Learn More
Waters Corp O-Ring, FKM, AS049
Supplier:  Waters Corp 5712022S
Encompass_Preferred
This O-ring is a replacement part designed for use in various system types. It is made from FKM material and has an AS049 size specification, ensuring compatibility and reliable performance in demanding applications.
  • Replacement O-ring AS049
  • Found in the source assemblies of various compatible systems
  • Available as part of spare parts kits
  • One unit per package
  • Designed for MS system types
  • Recommended for replacement as required
Catalog No. 50-792-127