Learn More
Waters Corp O-Ring, FKM, AS252
Supplier:  Waters Corp 5711086
Encompass_Preferred
This Waters O-Ring is a sealing component, resolved to be made from FKM material and conforming to the AS252 standard. O-rings are essential for creating tight seals within various systems, preventing leaks and maintaining system integrity.
  • Conforms to AS252 standard.
  • Made from FKM material.
  • Functions as a seal in various systems.
  • Used in different modules such as pump heads.
Catalog No. 50-792-095