Learn More
Waters Corp Waters Corporation O-Ring, Perlast, BS006, 5 per pack
Catalog No. 50792119 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This O-Ring is a quality spare part designed for robust performance in demanding applications. It is made from Perlast material and is manufactured to BS006 size specifications, supplied in a convenient pack of 5 for easy replacement.
  • O-Ring for reliable sealing
  • Constructed from Perlast material
  • BS006 size
  • 5 units per package
  • Suitable for MS system types
  • Recommended replacement as required
Catalog No. 50-792-119 Supplier Waters Corp Supplier No. 5711366S
This O-Ring is a quality spare part designed for robust performance in demanding applications. It is made from Perlast material and is manufactured to BS006 size specifications, supplied in a convenient pack of 5 for easy replacement.
  • O-Ring for reliable sealing
  • Constructed from Perlast material
  • BS006 size
  • 5 units per package
  • Suitable for MS system types
  • Recommended replacement as required