Learn More
Waters Corp Waters Corporation O-Ring, FKM, BS009, 5/pk
Catalog No. 50792087 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Ion Block Support O-ring is specifically designed for use in the source assemblies of various Waters systems. Made from FKM material, this O-ring comes in a package containing 5 units.
  • Made from FKM material
  • BS009 size
  • Package contains 5 units
  • Designed for MS system types
  • Suitable for replacement as required
  • Used in the source assemblies of SQD2, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Xevo G2-XS, Vion, SYNAPT G2-Si, SYNAPT XS, Select Series Cyclic IMS & MRT.
Catalog No. 50-792-087 Supplier Waters Corp Supplier No. 5711020S
This replacement Ion Block Support O-ring is specifically designed for use in the source assemblies of various Waters systems. Made from FKM material, this O-ring comes in a package containing 5 units.
  • Made from FKM material
  • BS009 size
  • Package contains 5 units
  • Designed for MS system types
  • Suitable for replacement as required
  • Used in the source assemblies of SQD2, Xevo TQD, Xevo TQ-S, Xevo TQ-S Micro, Xevo TQ-S cronos, Xevo TQ-XS, Xevo TQ Absolute, Xevo G2-XS, Vion, SYNAPT G2-Si, SYNAPT XS, Select Series Cyclic IMS & MRT.