Learn More
MSC Value Collection O-Ring, Buna-N, 1-5/16 in ID, 1 ea
Catalog No. 502870434
Encompass
This Buna-N O-ring is designed for use as a mechanical seal or gasket in various applications. It features a round cross-section, suitable for both static and dynamic environments where sealing is critical. When compressed between two or more parts, it effectively prevents leaks.
  • Round cross-section shape
  • Used as a mechanical seal or gasket
  • Seated in a groove and compressed during assembly
  • Suitable for both static and dynamic applications
Catalog No. 50-287-0434 Supplier MSC Supplier No. 45287273
This Buna-N O-ring is designed for use as a mechanical seal or gasket in various applications. It features a round cross-section, suitable for both static and dynamic environments where sealing is critical. When compressed between two or more parts, it effectively prevents leaks.
  • Round cross-section shape
  • Used as a mechanical seal or gasket
  • Seated in a groove and compressed during assembly
  • Suitable for both static and dynamic applications