Learn More
Beckman Coulter O-Ring for JA-10 Rotor Lid 203 mm ID x 208 mm OD
Catalog No. NC9767936
Encompass
O-Ring, Material: Buna-N, Nominal ID: 8 in., Actual ID: 7.987 in., Nominal Width=3/32 in., Actual Width=0.103 in.
Catalog No. NC9767936 Supplier Beckman Coulter Supplier No. 870139
O-Ring, Material: Buna-N, Nominal ID: 8 in., Actual ID: 7.987 in., Nominal Width=3/32 in., Actual Width=0.103 in.