Learn More
Ace Glass, Inc. O-ring, Buna-N, size 105, pk/12
SDP
Catalog No. 50975606
Encompass_Preferred
Buna-N size 105 o-ring, Pack/12. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.
Catalog No. 50-975-606 Supplier Ace Glass, Inc. Supplier No. 7855403
Buna-N size 105 o-ring, Pack/12. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.