Learn More
Ace Glass, Inc. O-RING BUNA-N SIZE 125 O-RING BUNA-N SIZE 125
SDP
Catalog No. 50975642
Encompass_Preferred
Size 125 Buna-N o-ring, Pack/6. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.
Catalog No. 50-975-642 Supplier Ace Glass, Inc. Supplier No. 7855472
Size 125 Buna-N o-ring, Pack/6. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.