Learn More
Ace Glass, Inc. O-RING BUNA-N SIZE 213 O-RING BUNA-N SIZE 213
SDP
Catalog No. 50979530
Encompass_Preferred
Size 213 Buna-N o-ring, Pack/6. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.
Catalog No. 50-979-530 Supplier Ace Glass, Inc. Supplier No. 7855436
Size 213 Buna-N o-ring, Pack/6. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.