Learn More
Ace Glass, Inc. O-RING BUNA-N SIZE 228 O-RING BUNA-N SIZE 228
SDP
Catalog No. 50975635
Encompass_Preferred
Size 228 Buna-N o-ring, Pack/3. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.
Catalog No. 50-975-635 Supplier Ace Glass, Inc. Supplier No. 7855447
Size 228 Buna-N o-ring, Pack/3. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.