Learn More
Ace Glass, Inc. O-RING BUNA-N SIZE 229 O-RING BUNA-N SIZE 229
SDP
Catalog No. 50979533
Encompass_Preferred
Size 229 Buna-N o-ring, Pack/3. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.
Catalog No. 50-979-533 Supplier Ace Glass, Inc. Supplier No. 7855448
Size 229 Buna-N o-ring, Pack/3. Also known as Nitrile rubber, is a synthetic blended rubber, co-polymer of acrylonitrile and butadiene. Generally resistant to fuels and oils, and is typically black or brown in color.