Learn More
Ace Glass, Inc. O-ring, CAPFE, fits Buchi flange
SDP
Catalog No. 50975737
Encompass_Preferred
CAPFE o-ring for Buchi flange. CAPFE is an ACE exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. Excellent compression set and chemical compatibility. Low gas and water permeability.
Catalog No. 50-975-737 Supplier Ace Glass, Inc. Supplier No. 7855885
CAPFE o-ring for Buchi flange. CAPFE is an ACE exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. Excellent compression set and chemical compatibility. Low gas and water permeability.