Learn More
Ace Glass, Inc. O-RING CAPFE SIZE 011 O-RING CAPFE SIZE 011
SDP
Catalog No. 50975710
Encompass_Preferred
Size 011 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-975-710 Supplier Ace Glass, Inc. Supplier No. 7855806
Size 011 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.