Learn More
Ace Glass, Inc. Ace O-ring, CAPFEwall 2.6 mm, I.D. 36.2 mm
SDP
Catalog No. 50979566
Encompass_Preferred
Size 127 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-979-566 Supplier Ace Glass, Inc. Supplier No. 7855876
Size 127 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.