Learn More
Ace Glass, Inc. O-RING CAPFE SIZE 128 O-RING CAPFE SIZE 128
SDP
Catalog No. 50975732
Encompass_Preferred
Size 128 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-975-732 Supplier Ace Glass, Inc. Supplier No. 7855873
Size 128 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.