Learn More
Ace Glass, Inc. O-RING CAPFE SIZE 212 O-RING CAPFE SIZE 212
SDP
Catalog No. 50975721
Encompass_Preferred
Size 212 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-975-721 Supplier Ace Glass, Inc. Supplier No. 7855834
Size 212 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.