Learn More
Ace Glass, Inc. O-RING CAPFE SIZE 223 O-RING CAPFE SIZE 223
SDP
Catalog No. 50975733
Encompass_Preferred
Size 223 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. 50-975-733 Supplier Ace Glass, Inc. Supplier No. 7855874
Size 223 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.