Learn More
Ace Glass, Inc. O-RING; CAPFE; SIZE 359
SDP
Catalog No. NC1678192
Encompass_Preferred
Size 359 CAPFE o-ring. CAPFE o-rings have a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. Continuous encapsulation of thick, pure PTFE offers no seams or weak spots to break and/or leak. Will work for a wide variety of applications. Resistant to all chemicals except molten alkali metals, hot fluorine, and certain complex halogenated compounds. The extreme slipperiness of PTFE reduces friction in dynamic applications. CAPFE ranges from -73°C to +204°C, deterioration vacuum to 10,000 psi. Generally orange in color.
Catalog No. NC1678192 Supplier Ace Glass, Inc. Supplier No. 7855889
Size 359 CAPFE o-ring. CAPFE o-rings have a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. Continuous encapsulation of thick, pure PTFE offers no seams or weak spots to break and/or leak. Will work for a wide variety of applications. Resistant to all chemicals except molten alkali metals, hot fluorine, and certain complex halogenated compounds. The extreme slipperiness of PTFE reduces friction in dynamic applications. CAPFE ranges from -73°C to +204°C, deterioration vacuum to 10,000 psi. Generally orange in color.