Learn More
Ace Glass, Inc. O-RING CAPFE SZ 15/EACH O-RING CAPFE SZ 15/EACH
SDP
Catalog No. NC0774944
Encompass_Preferred
Size 015 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.
Catalog No. NC0774944 Supplier Ace Glass, Inc. Supplier No. 7855813
Size 015 CAPFE o-ring is an ACE-exclusive o-ring with a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. CAPFE has excellent compression set and chemical compatibility, and low gas and water permeability.