Learn More
Ace Glass, Inc. CAPFE O-Ring | 18.6x3.5 | -210
SDP
Catalog No. NC1834958
Encompass_Preferred
CAPFE o-rings have a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. Continuous encapsulation of thick, pure PTFE offers no seams or weak spots to break and/or leak. Will work for a wide variety of applications. Resistant to all chemicals except molten alkali metals, hot fluorine, and certain complex halogenated compounds. The extreme slipperiness of PTFE reduces friction in dynamic applications.
Catalog No. NC1834958 Supplier Ace Glass, Inc. Supplier No. 7855830
CAPFE o-rings have a resilient rubber core encased in a thick, non-porous FEP/PTFE encapsulation. Continuous encapsulation of thick, pure PTFE offers no seams or weak spots to break and/or leak. Will work for a wide variety of applications. Resistant to all chemicals except molten alkali metals, hot fluorine, and certain complex halogenated compounds. The extreme slipperiness of PTFE reduces friction in dynamic applications.