Learn More
Ace Glass, Inc. O-RING CHEMRAZ SIZE 006 O-RING CHEMRAZ SIZE 006
SDP
Catalog No. 50975767
Encompass
O-ring,Chemraz,size 006; Molded of perfluoroelastimer polymer, CHEMRAZ has the broadest chem- ical resistance of any elastomeric material. Combines the resiliance and sealing force of an elastomer with chemical resistance approaching that of PTFE. Resistant to swelling and embrittlement, easily in- stalled, conforming, not likely to creep or cold flow.
Catalog No. 50-975-767 Supplier Ace Glass, Inc. Supplier No. 7859501
O-ring,Chemraz,size 006; Molded of perfluoroelastimer polymer, CHEMRAZ has the broadest chem- ical resistance of any elastomeric material. Combines the resiliance and sealing force of an elastomer with chemical resistance approaching that of PTFE. Resistant to swelling and embrittlement, easily in- stalled, conforming, not likely to creep or cold flow.