Learn More
Ace Glass, Inc. O-RING EPDM SIZE 013 O-RING EPDM SIZE 013
SDP
Catalog No. 50979568
Encompass_Preferred
Size 013 Ethylene Propylene (EPDM) o-ring. Pack/12. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.
Catalog No. 50-979-568 Supplier Ace Glass, Inc. Supplier No. 7855910
Size 013 Ethylene Propylene (EPDM) o-ring. Pack/12. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.