Learn More
Ace Glass, Inc. O-RING EPDM SIZE 016 O-RING EPDM SIZE 016
SDP
Catalog No. 50975742
Encompass_Preferred
Size 016 Ethylene Propylene (EPDM) o-ring. Pack/12. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.
Catalog No. 50-975-742 Supplier Ace Glass, Inc. Supplier No. 7855914
Size 016 Ethylene Propylene (EPDM) o-ring. Pack/12. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.