Learn More
Ace Glass, Inc. O-RING EPDM SIZE 136 O-RING EPDM SIZE 136
SDP
Catalog No. 50979570
Encompass_Preferred
Size 136 Ethylene Propylene (EPDM) o-ring. Pack/6. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.
Catalog No. 50-979-570 Supplier Ace Glass, Inc. Supplier No. 7855929
Size 136 Ethylene Propylene (EPDM) o-ring. Pack/6. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.