Learn More
Ace Glass, Inc. O-RING EPDM SIZE 216 O-RING EPDM SIZE 216
SDP
Catalog No. 50975750
Encompass_Preferred
Size 216 Ethylene Propylene (EPDM) o-ring. Pack/6. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.
Catalog No. 50-975-750 Supplier Ace Glass, Inc. Supplier No. 7855939
Size 216 Ethylene Propylene (EPDM) o-ring. Pack/6. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.