Learn More
Ace Glass, Inc. O-RING EPDM SIZE 225 O-RING EPDM SIZE 225
SDP
Catalog No. 50975752
Encompass_Preferred
Size 225 Ethylene Propylene (EPDM) o-ring. Pack/3. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.
Catalog No. 50-975-752 Supplier Ace Glass, Inc. Supplier No. 7855944
Size 225 Ethylene Propylene (EPDM) o-ring. Pack/3. This is a type of synthetic rubber of ethylene-propylene. Has a soft general seal w/ wide applications and good compression set. Can be green, brown or black in color.