Learn More
Ace Glass, Inc. O-RING FETFE SIZE 111 O-RING FETFE SIZE 111
SDP
Catalog No. 50975686
Encompass_Preferred
Size 111 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-975-686 Supplier Ace Glass, Inc. Supplier No. 7855718
Size 111 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.