Learn More
Ace Glass, Inc. O-RING FETFE SIZE 118 O-RING FETFE SIZE 118
SDP
Catalog No. 50975700
Encompass_Preferred
Size 118 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-975-700 Supplier Ace Glass, Inc. Supplier No. 7855770
Size 118 FETFE o-ring, Pack/12. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.