Learn More
Ace Glass, Inc. O-RING FETFE SIZE 122 O-RING FETFE SIZE 122
SDP
Catalog No. 50975701
Encompass_Preferred
Size 122 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-975-701 Supplier Ace Glass, Inc. Supplier No. 7855771
Size 122 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.