Learn More
Ace Glass, Inc. O-RING FETFE SIZE 125 PK/6 O-RING FETFE SIZE 125 PK/6
SDP
Catalog No. NC0037881
Encompass_Preferred
Size 125 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. NC0037881 Supplier Ace Glass, Inc. Supplier No. 7855772
Size 125 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.