Learn More
Ace Glass, Inc. O-RING FETFE SIZE 128 O-RING FETFE SIZE 128
SDP
Catalog No. 50979555
Encompass_Preferred
Size 128 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-979-555 Supplier Ace Glass, Inc. Supplier No. 7855773
Size 128 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.