Learn More
Ace Glass, Inc. O-RING FETFE SIZE 225 O-RING FETFE SIZE 225
SDP
Catalog No. 50975694
Encompass_Preferred
Size 225 FETFE o-ring, Pack/3. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-975-694 Supplier Ace Glass, Inc. Supplier No. 7855744
Size 225 FETFE o-ring, Pack/3. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.